Sleeve protectors, Bizweld™, BZ11
Fournisseur: Portwest
BZ11NAR
PRTWBZ11NAREA 124 EUR InStock
PRTWBZ11NAR
Sleeve protectors, Bizweld™, BZ11
Tenue de travail
Designed to give maximum protection and comfort to the wearer, the Bizweld™ sleeves will provide protection in hazardous conditions.
  • Features HRC2 and UPF 50+
  • The elasticated cuffs ensure a flexible but comfortable fit
  • Must be worn with a suitable coverall or jacket/trouser combination
  • Weight: 330 g
The garments in the Bizweld™ flame resistant range have been specially designed to guarantee comfort, performance and safety whilst providing effective function at all times. This range has been rigorously tested to ensure total compliance with the latest international standards.
This industry leading range provides multi standard protection for hazardous environments. These state of the art products are the result of years of experience combined with advanced technology and market research. Commitment to the health, safety and comfort of the wearer can be seen in the wide range of products suitable for all climates and end uses.
Certifications: EN ISO 11612 (A1+A2, B1, C1, E1, F1); EN ISO 11611 Class 1 (A1+A2); NFPA® 70E; ASTM F1959/F1959M-12 (ATPV=11,2 Cal/cm² (HAF=80,4%)).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité