Aluminium Twist Lock Carabiner, FP32
Fournisseur: Portwest
FP32SIR
PRTWFP32SIREA 16.7 EUR InStock
PRTWFP32SIR
Aluminium Twist Lock Carabiner, FP32
Protections anti-chutes Mousquetons
This lightweight twist-lock carabiner features a 20 mm gate for securing a connecting device such as a fall arrest lanyard to an anchorage point. Breaking load:23 kN.
  • Aluminium
  • Max rated load 100 kg
  • 10 Year shelf life from manufacturing date outlined in product label
Assort range of fall protection equipment to be used with our other fall protection equipment.
Height safety is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas. Portwest offer a full range of height safety products for working at height.
Certifications: CE certified. EN 362:2004 Class B, UKCA marked.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité