Flame-retardant balaclava, anti-static, Modaflame FR18
Fournisseur: Portwest
FR18NAR
PRTWFR18NAREA 22.8 EUR InStock
PRTWFR18NAR
Flame-retardant balaclava, anti-static, Modaflame FR18
Tenue de travail
This baclava provides excellent warmth and protection the to head and neck areas. It is made from Modaflame™ knit, an inherently fire resistant yarn of 60% modacrylic, 39% cotton and 1% carbon fibre.
  • Ribbed fabric provides a comfortable fit
  • Strong and durable fabric
  • Inherent flame resistant properties will not diminish with repeated washing
The baclava is flame resistant as well as anti-static making it suitable for multiple environments.
Certifications: EN ISO 11612 (A1, B1, C1, F1), EN 1149-5, IEC 61482-2, IEC 61482-1-2 (Class 1), NFPA 70E and ASTM F1959/F1959M-12 EBT=4,3 CAL/CM (HAF=66%).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité