Steelite™ S2 Safety Boots, Slip-On, Mid
Fournisseur: Portwest
FW83WHR40
PRTWFW83WHR40EA 38.1 EUR InStock
PRTWFW83WHR40
Steelite™ S2 Safety Boots, Slip-On, Mid
Chaussures et bottes
Slip-on boot with padded collar for added support. Excellent fit for all-day comfort and a steel toecap to keep feet safe.
  • Wipe clean upper
  • Slip-on silhouette
  • Available in unisex sizing
  • Machine washable to 30 °C
  • Anti-static footwear
  • Energy absorbing seat region
  • SRC - Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel- and oil-resistant outsole
  • Mono density sole unit
The Steelite™ Slip-on safety boot features a self-cleaning outsole to prevent cross-contamination along with a water-resistant upper to prevent water penetration. These boots are specially designed for use in cleanroom, medical, and food industry environments.
Certifications: EN ISO 20345: 2011 S2. CE certified.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité