Full face mask, Helsinki P516
Fournisseur: Portwest
P516BLU
PRTWP516BLUEA 143 EUR InStock
PRTWP516BLU
Full face mask, Helsinki P516
Respirators Masques réutilisables Masques complets
The Helsinki full face mask is made with a soft thermoplastic rubber face seal and features a panoramic viewer.
  • Excellent peripheral vision with 82% visibility
  • Six attachment harness for a uniform seal
  • Adjustable strap fastening for flexible sizing
  • Designed to be worn in a tough work environment, light weight
  • Materials: Thermoplastic rubber, polycarbonate, silicone, ABS
The internal silicone face piece is equipped with two inhalation valves to creduce fogging and improve comfort. This mask features standard thread connection according to EN 148-1.
It can be used with all filters supporting the same standard (gas, particles or combined and motorised equipment).
Certifications: EN 148 -1; EN 136 Class 3.
Informations de commande: Filters are not included (sold separately).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité