Gas filters, P976
Fournisseur: Portwest
P976BKR
PRTWP976BKREA 139 EUR InStock
PRTWP976BKR
Gas filters, P976
Filtres Filtres à gaz
ABEK2P3 combination filters are anti-gas and anti-particles filter and equipped with a standard threaded connection and can be used on full face masks with the same connection.
  • Lightweight and comfortable
  • Individually packed for vending machines
Protects from organic and inorganic acid gases and vapours, sulphur dioxide, ammonia and its derivatives. Classified as a class 2 filter for gases and class 3 for particles, they can be used to protect against harmful substances, solvents, toxic particles and micro-organisms.
Certifications: EN 148 -1, EN 14387 Type ABEK Class 2, EN 14387 Type P Class 3
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité