Dissolution paddles, QLA
Fournisseur: QUALITY LAB ACCESSORIES
EPD016-02
QLAAEPD016-02EA 0 EUR InStock
QLAAEPD016-02
Dissolution paddles, QLA
Appareils de dissolution
QLA paddles are precision manufactured to limit rotational wobble to levels below USP (711) specifications.
  • Made of high quality 316 electropolished stainless steel, PTFE coated over 316 stainless steel or solid fluoropolymer
  • All paddles are laser marked with a unique serial number
The single piece molded Fluoropolymer paddles provide a seamless construction eliminating corrosion and contamination encountered with conventional three-piece assemblies.
Informations de commande: Other paddle shaft lengths are also available on request.
Information de livraison: All paddles are supplied with a Certificate of Conformance (COC) stating that they are USP compliant and that they meet machine manufacturer specifications. A Certificate of Analysis (COA) is available at an additional cost.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité