Loading dye solution
Fournisseur: Thermo Fisher Scientific
R0611
R0611EA 46.3 EUR InStock
R0611
Loading dye solution
Colorants Colorants pour gels d'electrophorèse
This gel loading dye contains two different dyes bromophenol blue and xylene cyanol FF for visual tracking of DNA migration during electrophoresis. The presence of glycerol ensures that the DNA in the ladder and sample forms a layer at the bottom of the well. The EDTA included in the solution binds divalent metal ions and inhibits metal-dependent nucleases.
  • Two-colour tracking of DNA migration during DNA electrophoresis
  • No DNA masking during gel exposure to UV light
  • EDTA binds divalent metal ions and inhibits metal dependent nucleases
Used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité