RNA loading dye (2X)
Fournisseur: Thermo Fisher Scientific
R0641
R0641EA 41.99 EUR InStock
R0641
RNA loading dye (2X)
Colorants Colorants pour gels d'electrophorèse
In most denaturing agarose gel systems, bromophenol blue migrates slightly faster than human 5S rRNA, whereas xylene cyanol FF migrates slightly slower than 18S rRNA. RNA loading dye contains the denaturing agent formamide, which allows RNA fragments to separate according to size even during non-denaturing electrophoresis. Formamide also stabilises RNA.
  • Two-colour tracking of DNA, RNA migration during electrophoresis
  • No DNA, RNA masking during gel exposure to UV light
  • Denaturation of DNA, RNA due to the presence of formamide
Recommended for preparation of RNA samples for electrophoresis on agarose or polyacrylamide gels.
Informations de commande: Contains the tracking dyes bromophenol blue and xylene cyanol FF, as well as the intercalating dye ethidium bromide.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité