Ionselective Electrode, Ca for Blood Gas Analysers
Fournisseur: RADIOMETER CLINICAL PRODUCTS
945-616
RADC945-616EA 986.18 EUR InStock
RADC945-616
Ionselective Electrode, Ca for Blood Gas Analysers
Électrodes Électrodes ISE
The Ca electrode (E733) is an ionselective electrode whose sensing element is a PVC membrane containing a calcium-neutral ion carrier. The ionsensitive membrane is covered with a cellophane membrane in order to protect it from the samples.
The electrolyte has a constant and known concentration of calcium ions. When a sample is brought in contact with the electrode, a potential develops across the PVC and cellophane membranes.
The potential depends on the difference between the calcium (more precisely, activity) in the electrolyte and the sample. If the cCa²⁺ in both solutions is the same, the potential across the electrode tip will be 0 V.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité