Ion selective Electrode, Na for Blood Gas Analysers
Fournisseur: RADIOMETER CLINICAL PRODUCTS
945-618
RADC945-618EA 986.18 EUR InStock
RADC945-618
Ion selective Electrode, Na for Blood Gas Analysers
Électrodes Électrodes ISE
The Na electrode (E755) is an ionselective electrode whose sensing element is a Na⁺ -sensitive ceramic pin is contained in the tip of the jacket.
The electrolyte has a constant and known concentration of sodium ions. When a sample is brought in contact with the electrode, a potential develops across the ceramic pin.
The potential depends on the difference between the sodium (more precisely, activity) in the electrolyte and the sample. If the cNa⁺ in both solutions is the same, the potential across the electrode tip will be 0 V.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité