Single channel pipettes, electronic, variable volume, E4™ XLS™
Fournisseur: RAININ
17012312 17012313 17012314 17012353 17012354 17014486 17014483 17014490 17014494 17014484 17014487 17014482 17014488 17014489
RAIN17012312EA 0 EUR InStock
RAIN17012312 RAIN17012313 RAIN17012314 RAIN17012353 RAIN17012354 RAIN17014486 RAIN17014483 RAIN17014490 RAIN17014494 RAIN17014484 RAIN17014487 RAIN17014482 RAIN17014488 RAIN17014489
Single channel pipettes, electronic, variable volume, E4™ XLS™
Pipettes Micropipettes
The E4™ XLS™ is an electronic pipette that can be upgraded and expanded right from the PC. Available as standard versions or with LTS LiteTouch System for lower ejection forces.
  • Large screen, joystick control and carousel menus speed access to all modes and options
  • True Manual™ pipetting mode permits continuous Real Time control of pipetting speed and direction, combined with precise volume measurement
  • Independent control of aspirate, dispense and mix speeds for challenging liquids
  • Blowout and automatic dispensing options for precision pipetting protocols
  • Ergonomic fingerhook and joystick provide maximum comfort
Standard functions include basic pipetting, multi-dispensing, and mixing. Advanced functions include sequential volume programming, fixed volume, titration, dilution and reverse mode pipetting.
Information de livraison: Supplied with Li-Ion battery and wall power supply. An optional charging stand is available separately.
Commander maintenant
COMMANDE
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité