EDTA Standard Solution 0,05 M
Fournisseur: REAGECON
CP70051
REGECP70051EA 0 EUR InStock
REGECP70051
EDTA Standard Solution 0,05 M
Étalons Étalons pour l'analyse environnementale
Reagecon's EDTA 0,05 M is a fully factorised, high purity, stable product, developed and tested for titrations. This is an analytical volumetric solution, also called titrant, standard titrant or standard solution. It is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete. The product may be used for manual titrations, or more commonly automatic titrations also called potentiometric titration, to determine the end points by detection of any of several properties as detailed in several ChP monographs.
Certifications: According to Chinese Pharmacopoeia (ChP).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité