System Suitability Set to USP for Total Organic Carbon (TOC) Suitable for Use with Anatel A643 Analyser
Fournisseur: REAGECON
ISTOC1265
REGEISTOC1265EA 647 EUR InStock
REGEISTOC1265
System Suitability Set to USP for Total Organic Carbon (TOC) Suitable for Use with Anatel A643 Analyser
Étalons Étalons pour l'analyse environnementale
Reagecon's system suitability set to from USP for Total Organic Carbon (TOC) is a product of extremely high specification and purity. The product is produced from ultra pure raw materials including water that is prepared by a special proprietary process. The product is presented in bottles that are manufactured, cleaned and leached, by a special proprietary process specifically for the product.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité