Gasoline Calibration Standard (2 Compound Mix) in Petroleum Ether
Fournisseur: REAGECON
REGASCAL-8-250
REGEREGASCAL-8-250EA 920 EUR InStock
REGEREGASCAL-8-250
Gasoline Calibration Standard (2 Compound Mix) in Petroleum Ether
Étalons Étalons pour l'industrie pétrolière
Gasoline calibration standard (two compound mix) in petroleum ether is a ready to use standard used in fuel analysis as either a calibrant or analytical control solution. This standard is used in testing for a range of aromatic and oxygenate compounds in gasoline in accordance with the ASTM D5845, using infrared (IR) instrumentation. The product can also be used to validate appropriate test methods or qualify an instrument in a regulated industry. The product is QC tested on a high performance GC-MS.
  • Multi compound mix analyte
  • Mixed concentrations
  • Weight: 0,57 kg
  • Packing group II with class three
Certifications: Reagecon holds ISO 17025 accreditation (A2LA Ref: 6739.02) for calibration of laboratory balances.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité