Synonyms:
Salt of hartshorn
USP0605
REGEUSP0605EA
154
EUR
InStock
REGEUSP0605
Ammonium carbonate USP Test Solution (TS)
Ammonium carbonate
Reagecon's Ammonium Carbonate TS solution according to United States Pharmacopoeia (USP) is specified for use in individual United States Pharmacopoeia (USP) Monographs. It is NIST traceable and has guaranteed stability throughout its entire shelf life; even after opening the bottle.
Certifications: It is tested by an ISO 17025 Accredited Test Method (A2LA Ref: 6739.03).