Syringe adaptor kit for single-stage VOC regulator
Fournisseur: Restek
21118
REST21118EA 75.2 EUR InStock
REST21118
Syringe adaptor kit for single-stage VOC regulator
Matériel de chromatographie Chromatographie, Pièces de rechange
Used to withdraw sample from a high-pressure cylinder after pressure reduction through the high-purity VOC single-stage regulator.
  • High-purity regulator
  • Use with any syringe needle
Withdraws sample after pressure reduction.
Information de livraison: Kit contains one nickel-plated brass ¹/₄" NPT to female luer fitting, which can be used with an A-2 Luer syringe, and one stainless steel ¹/₄" NPT×¹/₈" compression fitting with septum (can be used with any syringe needle).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité