Hydrogen flashback arrestor
Fournisseur: Restek
21334
REST21334EA 644 EUR InStock
REST21334
Hydrogen flashback arrestor
Régulateurs de gaz
This hydrogen flashback arrestor is a special gas safety device, it cuts to stop the flame or reverse flow of gas back up into the equipment or supply line and it prevents the user and equipment from damage or explosions.
  • Valve cuts off gas supply in the event of a flashback
  • Can be reset and reused after flashback has occurred
  • Valve spring is made of 316 stainless steel
Maximum pressure: Hydrogen, 3,5 bar, temperature sensor: polyethylene
Certifications: Meets safety requirements of BS6158, EN730, ISO 5175, AS 4603 and are UL Listed 23Y5.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité