Gas trapping systems for LC/MS, Super Clean®
Fournisseur: Restek
22062
REST22062EA 1290 EUR InStock
REST22062
Gas trapping systems for LC/MS, Super Clean®
Filtres Filtres à gaz
The Super Clean® gas trapping system purifies nitrogen and is ideal for use in LC-MS systems. The two position base plate (installed in the gas line; standard base plate inlet/outlet fittings accept ¹/₄" tubing) allows cartridges to be exchanged without introducing impurities into the system. To meet the high flow needs of the LC-MS system, the activated charcoal filled cartridges are positioned and connected in parallel.
  • Up to 20 L of hydrocarbon free nitrogen per minute
  • Spring-loaded check valves allow cartridge replacement without contamination hazard
  • Estimated filter lifetime of 3 to 6 months
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité