Electronic Decapper and Crimper
Fournisseur: Restek
23237
REST23237EA 1530 EUR InStock
REST23237
Electronic Decapper and Crimper
Pinces à sertir et pinces à dessertir
This electronic crimpers and decappers have a rechargeable 6,4-volt battery installed. The large battery allows for quick, powerful operation and a faster charge cycle, minimising downtime. Charging the battery takes thirty minutes to two hours. Features a brushless motor and dual sleep mode options (light and deep) to extend battery life.
  • Hundreds of operations from one charge
  • Crisp OLED screen display lets you optimise crimping power for each cap type, monitor battery use, and view lifetime statistics
  • Large battery allows for faster charge cycle and two sleep mode options extend battery life
  • Easy to use; comfortable grip
The crisp, easy-to-read OLED screen display lets you monitor battery status, crimp setting, cycle results, and a scrolling error message at a glance. Crimp setting is the percent of power being used, and it can be controlled and optimised for different cap types. Access the settings menu for lifetime statistics, recent cycle log info, eight language options, and reset functions.
One-year warranty.
Information de livraison: All kits include a universal plug set for the battery charger.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité