Sample delivery system, Resprep®
Fournisseur: Restek
26250
REST26250EA 559 EUR InStock
REST26250
Sample delivery system, Resprep®
Matériel de chromatographie Chromatographie, Pièces de rechange
This sample delivery system is used to transfer large volumes of low viscosity samples directly from a bottle to a solid phase extraction cartridge or to a disk on a vacuum manifold system for extraction or cleanup.
  • Compatible with 1, 3, 6, and 15 ml SPE cartridges and Diskcover-47 extraction disk holder
  • Tested to pH 1 to ensure no contaminants leach from system
  • Specified in EPA drinking water methods
Information de livraison: Each unit consists of six PTFE transfer lines with a stainless steel weight on one end and a colour coded screw fitting with PET adapter on the opposite end.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité