Trident LC Column Protection System
Fournisseur: Restek
27473
REST27473EA 491 EUR InStock
REST27473
Trident LC Column Protection System
Colonnes pour chromatographie
Trident LC column protection system is available in three levels of protection such as filter only, cartridge only, or filter and cartridge.
  • Easy-to-remove cap frit simplifies filter replacement
  • Low-dead-volume design has negligible effect on chromatography
  • Direct connection eliminates tubing and connectors that increase system volume and leak potential
The durable metal tip with replaceable PEEK ferrule means easy installation onto column without tools. Improved thread design and materials create an optimal seal that releases and reseals easily, allowing multiple installations without galling and binding.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité