PAL SPME fibers
Fournisseur: Restek
27480.1
REST27480.1EA 0 EUR InStock
REST27480.1
PAL SPME fibers
Matériel de chromatographie Chromatographie, Pièces de rechange
SPME automated sample preparation reduces sample handling and solvent consumption.
  • Suitable for a wide range of analyte chemistries and sample matrices
  • Reliable performance meets or exceeds other brands
  • Robust aluminum hub is more durable than plastic
  • Optimized for PAL system autosamplers and compatible with most GC inlets
All PAL SPME fibers are 10 mm in length and are housed in a 23-gauge needle.
Solid phase microextraction (SPME) is an automated sample preparation technique that reduces sample handling and solvent consumption. SPME technologyMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité