Weathered gasoline kit #2
Fournisseur: Restek
30437
REST30437EA 251 EUR InStock
REST30437
Weathered gasoline kit #2
Étalons Étalons pour l'industrie pétrolière
These solutions are prepared from a single-source (one-refinery) product. The weathered materials indicate the percent weight loss from the original material. Samples of regular- and premium-grade unleaded gasoline were collected, then blended in equal volumes.
Contains: 30096: Unleaded Gasoline (ASTM Class 2 Accelerant); Unleaded gasoline: unweathered (8006-61-9);30097: Unleaded Gasoline (ASTM Class 2 Accelerant); Unleaded gasoline: 25% weathered (8006-61-9);30098: Unleaded Gasoline (ASTM Class 2 Accelerant); Unleaded gasoline: 50% weathered (8006-61-9);30099: Unleaded Gasoline (ASTM Class 2 Accelerant); Unleaded gasoline: 75% weathered (8006-61-9);30436: Unleaded Gasoline (ASTM Class 2 Accelerant); Unleaded gasoline: 99% weathered (8006-61-9).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité