DHA Olefins standard
Fournisseur: Restek
30727
REST30727EA 881 EUR InStock
REST30727
DHA Olefins standard
Étalons Étalons organiques
This designed for the determination of the individual hydrocarbons present in spark ignition engine fuels, as well as fuel blends containing oxygenates such as methyl tert-butyl ether, ethyl tert-butyl ether, tert-butanol and ethanol.
  • 26 components
This standard is produced for refineries performing Detailed Hydrocarbon Analysis (DHA) of crude oil feedstocks and fuels. It helps with calibrating complex hydrocarbon analyses and provides the greatest number of gravimetrically determined values for quantitative calibration.
Certifications: ASTM Methods D6729, D6730, and D6733 (Determination of individual components in spark ignition engine fuels)
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité