MOSH/MOAH Retention Time Standard
Fournisseur: Restek
31076
REST31076EA 88.8 EUR InStock
REST31076
MOSH/MOAH Retention Time Standard
Étalons Étalons de chromatographie gazeuse
Developed specifically for using LC coupled with GC-FID to accurately determine hydrocarbons found in mineral oils, which can migrate from processing equipment and packaging into food.
  • Weight: 0,05 lbs.
  • W×L×H: 1,25×1,25×3,5"
  • Shipping condition: Ambient
This 10 component mix contains both mineral oil saturated hydrocarbon (MOSH) and mineral oil aromatic hydrocarbon (MOAH) markers that are used to accurately identify GC retention time limits for integration.
It contains 10 components: (C10) n-Decane (124-18-5), (C11) n-Undecane (1120-21-4), (C13) n-Tridecane (629-50-5), (C16) n-Hexadecane (544-76-3), (C20) n-Eicosane (112-95-8), (C24) n-Tetracosane (646-31-1), (C25) n-Pentacosane (629-99-2), (C35) n-Pentatriacontane (630-07-9), (C40) n-Tetracontane (4181-95-7), (C50) n-Pentacontane (6596-40-3).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité