TNRCC 1005 retention time marker mix (9 components)
Fournisseur: Restek
31698
REST31698EA 67.7 EUR InStock
REST31698
TNRCC 1005 retention time marker mix (9 components)
Étalons Étalons pour l'analyse environnementale
This TNRCC 1005 retention time marker mix used during initial sample screening to determine retention time windows for each petroleum product.
  • Easily determine the retention time window for each boiling point range
  • Prepared in n-pentane according to EPA requirements
Gasoline generally elutes in the window from C6 to C10 (or C12), and diesel fuel from C10 (or C12) to C24 (or C28). Retention above C24 (or C28) indicates oil or lubricant contamination.
It has four components: n-Hexane, n-Dodecane, n-Octacosane, n-Pentatriacontane.
Shelf life 6 to 85 months.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité