Carbohydrate LC performance check mix (5 components)
Fournisseur: Restek
31809
REST31809EA 67.7 EUR InStock
REST31809
Carbohydrate LC performance check mix (5 components)
Étalons Étalons de chromatographie gazeuse
This carbohydrate LC performance check mix consist of five components which performance qualification (PQ) determines the precision of the LC system.
  • For simple sugar analysis by LC with refractive index/light scattering detection
  • Use with a silica column with amino functionality, as recommended by AOAC International
  • Use as a calibration standard for AACC Method 80-04
  • Offered dry for enhanced stability
Check mix for LC-RI consists of five simple sugars in varied concentrations: Glucose (2,1 mg), Fructose (2,0 mg), Lactose (4,4 mg), Maltose (4,5 mg), Sucrose (4,0 mg).
Performance qualification (PQ) determines the precision of the LC system. Our performance check mix for LC-RI consists of five simple sugars in varied concentrations. We prepare the reference material in water, dehydrate it, and package it dry for enhanced stability.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité