Fortification Mix (7 components)
Fournisseur: Restek
31813
REST31813EA 103 EUR InStock
REST31813
Fortification Mix (7 components)
Étalons Étalons organiques
EPA Contract Lab Program (CLP) Statement of Work (SOW) OLC03.2 is an analytical method for the detection of low-concentration volatile, semivolatile, and pesticide/Aroclor organics in aqueous samples using deuterated monitoring compounds (DMCs) as sample-by-sample accuracy indicators. Restek supports this method with a convenient VOA MegaMix (with m- and p-xylene isomers at half the concentration of other components), a deuterated monitoring compounds (DMCs) mix, a fortification mix for all compounds requiring calibration at higher concentration, and single-compound solutions of
3,3'-dichlorobenzidine, and 4-chloroaniline.
This fortification mix contains: 4,6-Dinitro-2-methylphenol (Dinitro-o-cresol), 2,4-Dinitrophenol, 2-Nitroaniline, 3-Nitroaniline, 4-Nitroaniline, 4-Nitrophenol, 2,4,5-Trichlorophenol.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité