31971
REST31971EA
4160
EUR
InStock
REST31971
LC multiresidue pesticide kit
Étalons
Étalons organiques
Accurately detect and quantify pesticides of global food safety concern in a wide range of fruits, vegetables, and other commodities by LC-MS/MS.
- Quantitatively tested to confirm composition, detailed support documentation provided
Formulated and grouped for maximum long-term stability* and well-balanced chromatographic performance.
Optimised multiresidue pesticide method is offered free of charge and downloadable XLS file includes conditions and transition tables.
Contains 1 ml each of the following mixtures, LC multiresidue pesticide standard #1, LC multiresidue pesticide standard #2, LC multiresidue pesticide standard #3, LC multiresidue pesticide standard #4, LC multiresidue pesticide standard #5, LC multiresidue pesticide standard #6, LC multiresidue pesticide standard #7, LC multiresidue pesticide standard #8, LC multiresidue pesticide standard #9 and LC multiresidue pesticide standard #10.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Informations de commande: The complete kit contains 204 compounds of interest, covering many LC-determined pesticides listed by government agencies, individual ampoules also sold separately.
* NOTE: When combining many compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 204 compounds involved, and then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analysing each mix separately to ensure accurate results for every compound.