FAPAS series 9 OP pesticide mix 1 (10 components)
Fournisseur: Restek
32413
REST32413EA 0 EUR InStock
REST32413
FAPAS series 9 OP pesticide mix 1 (10 components)
Étalons Étalons organiques
Laboratories testing food quality and safety are encouraged to routinely perform proficiency tests. Proficiency testing is an external check of quality. It provides an independent and unbiased assessment of the performance of all aspects of the laboratory, both human and hardware.
  • External check of quality for laboratories performing food testing
  • Ensures accurate proficiency testing
  • Equal concentration of all compounds and suitable for GC-FPD, GC-NPD, and GC-MS analysis
Each participating laboratory is encouraged to use its normal analytical method, thereby simulating the testing of a routine laboratory sample as closely as possible.
Contains ten components, those are chlorpyriphos, chlorpyriphos-methyl, diazinon, dichlorvos , etrimphos, fenitrothion, malathion, methacriphos, phosphamidon and pirimiphos-methyl.
Certifications: Certified reference material (CRM) manufactured and QC-tested in Restek’s ISO-accredited labs—satisfy your ISO requirements.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité