FAPAS series 5 OC pesticide mix 2 (19 components)
Fournisseur: Restek
32414
REST32414EA 121 EUR InStock
REST32414
FAPAS series 5 OC pesticide mix 2 (19 components)
Étalons Étalons organiques
Laboratories testing food quality and safety are encouraged to routinely perform proficiency tests. Proficiency testing is an external check of quality. It provides an independent and unbiased assessment of the performance of all aspects of the laboratory, both human and hardware.
  • External check of quality for laboratories performing food testing
  • Ensures accurate proficiency testing
  • Varied concentrations and suitable for GC-ECD analysis
Each participating laboratory is encouraged to use its normal analytical method, thereby simulating the testing of a routine laboratory sample as closely as possible.
Contains 19 components, those are aldrin, α-bhc, β-bhc, γ-bhc , cis-chlordane, trans-chlordane, 4,4'-ddd, 4,4'-dde, 2,4'-ddt, 4,4'-ddt, dieldrin, α-endosulfan, β-endosulfan, endosulfan sulfate, endrin, heptachlor and heptachlor epoxide.
Certifications: Certified reference material (CRM) manufactured and QC-tested in Restek’s ISO-accredited labs—satisfy your ISO requirements.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité