GC multi-residue pesticide standard #8 (24 components)
Fournisseur: Restek
32570
REST32570EA 355 EUR InStock
REST32570
GC multi-residue pesticide standard #8 (24 components)
Étalons Étalons organiques
The multiresidue accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
  • Quantitatively tested to confirm composition, detailed support documentation provided
  • Formulated and grouped for maximum long-term stability
The multiresidue pesticide standard consist of 24 components bromfenvinfos-methyl, bromfenvinphos, bromophos ethyl, bromophos methyl, carbophenothion, chlorfenvinphos, chlorthiophos, coumaphos, edifenphos, ethion, fenamiphos, fenchlorphos (ronnel), fenthion, iodofenphos, leptophos, malathion, methacrifos, profenofos, prothiofos, sulphotepp, sulphrofos, terbufos, tetrachlorvinfos, tolclofos-methyl.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité