BAC resolution control standard n-P (6 components)
Fournisseur: Restek
36010
REST36010EA 55.5 EUR InStock
REST36010
BAC resolution control standard n-P (6 components)
Étalons Étalons organiques
The mix is use to verify the retention time for each compound normally included in a blood alcohol test, and to verify that the compounds are resolved from and do not interfere with one another.
  • Includes 1-propanol internal standard
  • Intended for qualitative use only
It consists of acetaldehyde, acetone, ethanol (BAC), methanol, 1-propanol (n-propanol) and 2-propanol (isopropanol).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité