Force EXP Guard Column Cartridge
Fournisseur: Restek
963950252
REST963950252EA 488 EUR InStock
REST963950252
Force EXP Guard Column Cartridge
Colonnes pour chromatographie
Free-Turn architecture lets you change cartridges by hand without breaking inlet/outlet fluid connections, with no tools needed.
  • Titanium hybrid ferrules
  • High pressure seal
  • Auto-adjusting design
Patented titanium hybrid ferrules can be installed repeatedly without compromising high-pressure seal. Auto-adjusting design provides ZDV (zero dead volume) connection to any 10 to 32 female port. Guard column cartridges require EXP direct connect holder. Pair with EXP hand-tight fitting for tool-free installation. For use with 3 or 5 µm Force LC columns. For 1,8 µm Force columns, use a 0,2 µm UltraShield filter.
Informations sur les accessoires: EXP Direct Connect Holder is available separately.
Informations de commande: To help protect your investment and further extend the life of our already-rugged LC columns, Restek offers the patent-pending guard column hardware developed by Optimise Technologies. A Restek LC guard cartridge in an EXP direct connect holder is the ultimate in column protection.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité