Cyclone mill TWISTER
Fournisseur: Retsch GmbH
20.831.0001
RETS20.831.0001EA 7540 EUR InStock
RETS20.831.0001
Cyclone mill TWISTER
Broyeurs
The TWISTER is specially designed for the processing of foods and feeds for subsequent NIR analysis. The optimised form of rotor and grinding chamber generates an air jet which carries the ground sample through the integrated cyclone into the sample bottle.
  • Ideal for grinding feeds, grains, forage and other similar products
  • Three controlled speeds
  • Cyclone separator with 250 ml collecting bottle for quick extraction of sample
  • No cross contamination thanks to easy cleaning
  • Convenient operating panel
The rotor speed can be adjusted in three steps allowing for perfect adaptation to the sample requirements. Cleaning the mill is quick and easy as the air jet effects a complete discharge of the material from the grinding chamber.
The air jet prevents the material from heating up, thus preserving the moisture content.
Information de livraison: Twister is supplied complete with rotor, grinding ring, sieve inserts (1,0 mm and 2,0 mm) and 10 sample bottles 250 ml.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité