Tamiseuses analytiques AS 200 tap
Fournisseur: Retsch GmbH
30.025.0001 30.025.1001
RETS30.025.0001EA 4500 EUR InStock
RETS30.025.0001 RETS30.025.1001
Tamiseuses analytiques AS 200 tap
Agitateurs Tamiseurs
Analytical sieve shaker AS 200 tap is used in research and development, quality control of raw materials, interim and finished products, as well as in production monitoring.
  • Maintenance-free
  • Excellent separation efficiency even with short sieving times
  • Digital time setting
  • Easy operation, ergonomic design
  • Adjustable plate for sieve stack
Mechanical action is applied to the test sieves in two dimensions. The combination of horizontal circular motion with 280 oscillations and a vertical tapping motion with 150 taps per minute helps the particles to pass the sieve apertures. This combination of sieving motions simulates hand sieving.
The tapping motion of this shaker supports the sieve analysis of certain products such as activated carbon, abrasives, metal powder, spices and diamonds, as specified in the corresponding standards.
Oscillations and taps are independent of power frequency which makes analysis results comparable world wide. The optional software EasySieve® for control through RS232 serial interface helps for easy evaluation and documentation of results.
Certifications: Conforms to ISO 9001.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité