Racks for sliding drawer freezer, Matrix, Matrix 2D and Nunc CryoBank
Fournisseur: Thermo Scientific
1950815 1950799 1950789 1950819 1950787 1950641
HERA1950815EA 189 EUR InStock
HERA1950815 HERA1950799 HERA1950789 HERA1950819 HERA1950787 REVC1950641
Racks for sliding drawer freezer, Matrix, Matrix 2D and Nunc CryoBank
Portoirs Portoirs pour cryoboîtes
Maximise tube storage capacity per freezer with Thermo Scientific™ Sliding drawer racks for tubes (4 inner door freezers), a racking solution for four inner-door, ultra-low temperature freezers. Use in conjunction with Thermo Scientific™ dense storage vials.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Standard/deepwell microplate racks
  • Warranty varies by country
  • Material: Stainless steel
  • Colour: Silver
Thermo Scientific freezer racking and tubing solutions offer a convenient, streamlined process to maximise sample storage in your laboratory. Our racking solutions are designed to hold tubes ranging from 200 µl up to 5,0 ml, offering maximised sample storage capacity for both five and four inner door freezers.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité