PEEK tubing, Rheodyne®
Fournisseur: IDEX RHEODYNE
1532
RHEO1532EA 58.5 EUR InStock
RHEO1532
PEEK tubing, Rheodyne®
Tuyaux Tubulures à chromatographie
Polymer tubing, coloured.
  • Biocompatible, inert and easy to cut
  • Great for high pressure applications
  • Many sizes colour-coded for easier inner diameter identification
PEEK (polyetheretherketone) tubing is biocompatible, chemically inert and can be used in place of stainless steel tubing in most liquid analytical systems. Additionally, when the Ø is smaller than ¹/₁₆", PEEK tubing can be used in place of more traditional fused silica capillary tubing. PEEK tubing can be used with stainless steel nuts and ferrules, flangeless fittings, Super Flangeless™ fittings, or any Upchurch Scientific-brand universal Fingertight fitting.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité