Anti-Protein A Polyclonal Antibody
Fournisseur: Rockland Immunochemicals
200-901-077
ROCK200-901-077EA 566 EUR InStock
ROCK200-901-077
Anti-Protein A Polyclonal Antibody
Anticorps
Chicken anti-protein A antibody.
  • Purified IgY fraction
  • ELISA, western blot
Protein A is 56 kDA surface protein found in the cell wall of Staphylococcus aureus. Protein A has a high affinity to Human IgG, Mouse IgG and others and is therefore suitable for purification and detection of immunoglobulins.
Anti-PROTEIN A (CHICKEN) Antibody has been tested for use in ELISA and immunoblotting. Although not tested, this antibody may also be useful for neutralisation assays, immunohistochemistry, flow Cytometry, immunoprecipitation, immunodiffusion, conjugation and most immunological methods requiring high titter and specificity.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité