Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)
Fournisseur: Rockland Immunochemicals
606-1302
ROCK606-1302EA 308 EUR InStock
ROCK606-1302
Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)
Anticorps
Anti-Guinea Pig IgG whole molecule is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays. Specific conditions for reactivity and signal detection should be optimized by the end user.
Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity:More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité