Tween-Tris-buffered saline (TTBS), 10X concentrate, pH 7,5
Fournisseur: Rockland Immunochemicals
MB-013
ROCKMB-013EA 195 EUR InStock
ROCKMB-013
Tween-Tris-buffered saline (TTBS), 10X concentrate, pH 7,5
Tampons Tampons d'électrophorèse Tampons de lavage
Tween Tris buffered saline is suitable for multiple applications including biological diluent buffer for antibodies or other biologics. Also may be used as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA.
This product is a 10X concentrated stock solution and should be diluted appropriately with distilled, deionised water or equivalent to its final working concentration. This buffer consists of 1,0 M Tris HCl, 1,5 M sodium chloride and 0,1% (v/v) Tween-20 at a pH of 7,5. Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionised water.
10X TTBS solution was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Attention: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité