Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)
Fournisseur: Rockland Immunochemicals
MB-016
ROCKMB-016EA 165 EUR InStock
ROCKMB-016
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)
Buffers Tampons de clonage
This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Attention: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité