GET buffer
Fournisseur: Rockland Immunochemicals
MB-043
ROCKMB-043EA 159 EUR InStock
ROCKMB-043
GET buffer
Buffers Tampons de clonage
GET buffer (50 mM Glucose, 10 mM EDTA, 25 mM Tris HCl, pH 8,0) is used in effective plasmid miniprep.
This product is a 1X concentrated stock solution. No dilution is required. Meticulously prepared using ultra pure reagents dissolved in pharmaceutical grade deionised water.
This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité