Antibacterial Foam Soaps
Fournisseur: Rubbermaid Commercial Products
2167858 2167901 2167860 2167904
RUBB2167858EA 23.5 EUR InStock
RUBB2167858 RUBB2167901 RUBB2167860 RUBB2167904
Antibacterial Foam Soaps
Soin de la peau Savons et soins nettoyants
Antibacterial soap kills germs, which is ideal for facilities looking for added peace of mind.
  • Kills 99,99% of common germs; fully tested and proven to be bactericidal, virucidal, and yeasticidal
  • Broad spectrum activity disinfects hands in 30 seconds
  • Fragrance-free; non-tainting for food environments
  • Pleasant to use, rich foam formula is ideal for any location
This fast-acting, fragrance-free solution contains moisturising ingredients to help prevent skin dryness. The biodegradable formula supports environmental wellbeing. Controlled dispensingMore Product Information
Commander maintenant
COMMANDE
SPÉCIFICATIONS
PRODUITS COMPLEMENTAIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité