Ladles, USP Class VI, double-bagged
Fournisseur: Sampling Systems
9053H-50S
SAMP9053H-50SEA 172 EUR InStock
SAMP9053H-50S
Ladles, USP Class VI, double-bagged
Pelles
These sterile HDPE ladles are great for taking fast liquid samples. The entire range have a slim line design so they can work through 2" bungs. They are commonly used for taking liquid samples from tubs, containers, bins etc.
  • Double-bagged
  • Produced in Class 100,000 cleanroom, packed in Class 10,000 cleanroom
The ladles are supplied double bagged as part of our SteriWare Ultra range. The box is also lined with a further bag for extra protection.
Certifications: USP Class VI Medical grade polymer is FDA and EC compliant, tested to USP661. Certified BSE/TSE free.
Conditionnement: Supplied in a lined box.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité