Stainless Steel Tray
Fournisseur: Sampling Systems
A236-500
SAMPA236-500EA 193 EUR InStock
SAMPA236-500
Stainless Steel Tray
Plateaux/Récipients/Pots Plateaux et récipients de laboratoire
Designed for heavy-duty use in pharmaceutical, chemical, and food facilities.
  • Made from high-quality 316L stainless steel
  • Mirror polish finish and radiused corners for easy cleaning
  • Available from stock
  • Fast delivery
316L stainless steel trays are perfect for heavy-duty use across a range of industries, including food and pharmaceutical. Design features in line with GMP guidelines ensure they are easy to both use and clean.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité