Stainless Steel Buckets, Graduated
Fournisseur: Sampling Systems
A264-15 A264-10 A264-10L A264-12 A264-12L A264-15L
SAMPA264-15EA 181 EUR InStock
SAMPA264-15 300-0649 300-0650 300-0651 300-0652 300-0653
Stainless Steel Buckets, Graduated
Godets
Stainless steel, graduated buckets are ideal for use in food, chemical, and general industry.
  • 304 stainless steel construction
  • Skirtband gives extra protection to the base
  • Robust design
  • Easy to use
Graduated stainless steel buckets are made from heavy gauge stainless steel and are very durable. They are ideal for general-purpose factory work. The swing handle allows them to be lifted easily. Optional lids are available separately.
The 10,12 and 15 litre buckets feature a skirt band around the base. This provides extra protection to the base andMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité