Stainless Steel Tweezers
Fournisseur: Sampling Systems
A394-100 A394-200 A394-300 A394-400
SAMPA394-100EA 4.2 EUR InStock
SAMPA394-100 SAMPA394-200 SAMPA394-300 SAMPA394-400
Stainless Steel Tweezers
Pinces
These stainless steel tweezers are perfect for use in laboratory and production environments.
  • Made from 304 stainless steel
  • Available in a range of different lengths
  • Robust design
  • Useful for handling intricate items such as tissues
This range of stainless steel tweezers are perfect for a variety of applications across a wide range of industries including pharmaceutical. They are great for handling small items such as tissues and vials and are made from medical grade stainless steel.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité