X-ray and metal detectable long handle scoops
Fournisseur: Sampling Systems
M1040B-100
SAMPM1040B-100EA 28.6 EUR InStock
SAMPM1040B-100
X-ray and metal detectable long handle scoops
Pelles
These long handled metal detectable scoops are perfect for both sampling and dispensing when working with powders and granules.
  • Robust PP construction gives excellent chemical resistance to many products
  • Supplied Individually bagged and produced in a cleanroom
  • Full batch traceability
The long handle and the bright blue colour make the scoops easy to spot if they get left behind. They are both X-ray and metal detectable.
To ensure the cleanliness of the scoops they are manufactured and packed in a Cleanroom. The robust nature of these scoops means that they can give many years of service or for more demanding applications they can be used as a single use disposable product straight from the bag.
Certifications: These scoops conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité